Project Star Teacher's Sampler and Cardboard Spectrometer
Supplier: SCIENCE INTERACTIVE GROUP
654-0120 654-0110
470350-126EA 401.49 CAD
470350-126 470350-098
Project Star Teacher's Sampler and Cardboard Spectrometer
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
This sampler of project STAR materials provides hours of astronomy activities for your classroom.
Delivery information: The sampler includes (one of each): do-it-yourself star finder kit; celestial sphere kit; sun tracking hemisphere kit; 3-D constellation kit; spectrometer kit (cardboard); refracting telescope kit (with tubes); light measurement and stellar distance kit; holographic diffraction grating - mounted, 35 mm glass slide; and where we are in space and time activity book.