Dropping Bottle
Supplier: United Scientific Supplies
PDB007PK12 PDB010PK12 PDB015PK12 PDB030PK12 PDB060PK12 PDB125PK12
470349-808PK 8.2 CAD
470349-808 470349-752 470349-690 470349-730 470349-692 470349-782
Dropping Bottle
Bottles Dropping Bottles
Dispense solutions accurately, one drop at a time.
  • Ideal for storing and dispensing small volumes
  • Translucent design
  • LDPE with leakproof PP cap
  • BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.