Value Plastics® Fittings, Y-Adapter, Hose Barb Union, PVDF
Supplier: VALUE PLASTICS TE
N670-J2A (100PK) MFLX40617-70BLK Y260-J2A (100PK)
MFLX40617-21BLKEA 232.26 CAD
MFLX40617-21BLK MFLX40617-70BLK MFLX40617-71BLK
Value Plastics® Fittings, Y-Adapter, Hose Barb Union, PVDF
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, or food processing.