AI Bacteria Analysis Slides
Supplier: Apacor
255000 255001
76670-670PK 36.97 CAD
76670-670 76670-672
AI Bacteria Analysis Slides
Slides Microscope Slides
Fiducial printed glass microscope slides for AI image analysis.
  • Printed fiducial design
  • White glass
  • Cut edges
Slides are available in two formats, with either a circle center or square center.