Alsto JC3020 Jaw Crusher
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
402245 402246
76638-294EA 229108.15 CAD
76638-294 76638-296
Alsto JC3020 Jaw Crusher
Mills
The Alsto JC3020 is suitable for either laboratory use or plant applications. Rugged construction and high reduction capabilities together with low maintenance requirements make it outstanding for a large variety of applications.
  • Large input particle size capability
  • 300×200 mm jaw opening allows for lump sizes of up to 170 mm
  • 25 mm product size allows for fast, efficient reduction with a high throughput of 3000 kg/hr
  • Simple and effective single toggle design reduces maintenance and repair time
  • Dust extraction points at feed hopper and crusher body and full drive train guarding ensures operator safety
  • Easily accessible sample drawer
  • Fast and easy jaw gap adjustment, ensuring positive control of product sizing
  • Hard wearing jaw plates and side liners
The Alsto JC3020 jaw crusher is a large unit suitable for either laboratory use to process samples with larger particle sizes or plant applications. It’s rugged construction and high reduction capabilities together with low maintenance requirements make it an outstanding workhorse for a large variety of applications.