Alsto DP2000 Disc Pulveriser
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
402234 402237
76638-282EA 58098.2 CAD
76638-282 76638-284
Alsto DP2000 Disc Pulveriser
Mills
Simple, robust machine designed for fine grinding of a large variety of sample and ore types. One fixed and one rotating 200 mm grinding plate turning on a horizontal axis, with the sample drawer located at the bottom of the chamber.
  • A simple screw adjustment to control product sizing via plate gap setting
  • Powerful 4kW, 3 phase drive motor
  • Supplied as standard on a base plate suitable for bench/workstation mounting
  • Durable chilled cast iron disc plates for long life
The Alsto DP2000 Disc Pulveriser is a simple, robust machine designed for fineMore Product Information