Plasmid DNA Kit
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
DNF-940-0240 DNF-940-K0500
CA76645-364EA 602.85 CAD
CA76645-364 CA76645-276
Plasmid DNA Kit
Electrophoresis Systems Capillary Electrophoresis Systems
Quality control analysis of supercoiled and linear plasmid DNA for downstream applications.
  • Reliable sizing- optimized ladder for the accurate sizing of supercoiled plasmids
  • Concentration range - 0.1 to 1 ng/ul
  • Fast analysis - QC results in only 30 minutes
  • Accurate sizing - sizing range from 2,000 to 10,000 bp for supercoiled plasmids
The plasmid DNA kit is used in the analysis of supercoiled and linear plasmid DNA. The ladder provided with this kit is optimized for the accurate sizing and relative quantification of supercoiled plasmids between 2,000More Product Information