Small RNA Kits
Supplier: Agilent
DNF-262-0250 DNF-262-0250 DNF-470-0275 DNF-470-0275
CA76645-316EA 1350.32 CAD
CA76645-316 76645-054 76645-316 CA76645-054
Small RNA Kits
Electrophoresis Systems Capillary Electrophoresis Systems
Analysis of small RNA quality control including microRNA quantification.
  • Broad concentration range - The small RNA kit has a concentration range of 25 to 2500 pg/µl
  • Short analysis time - Analysis time in as little as 22 minutes
  • Reliable sizing - Sizing range from 15 to 200 nt ensures accurate and precise sizing of small RNA samples
  • Low sample volume - Only 2 µl of sample is required, minimizing sample loss to QC steps
  • Specific RNA analysis modes - ProSize data analysis software uses a specific small RNA analysis region function automatically calculating percent microRNA and quantification of microRNA and small RNA
  • Capillary array - Optimized for the 12 -capillary array
The small RNA kit focuses on microRNA and small RNA quality control. Analysis of small RNAs is essential for downstream applications such as small RNA NGS libraries. The small RNA ladder includes six fragments ranging from 15 to 200 nt, which provides accurate and precise quantification and sizing of small RNA and microRNA. Focusing on a narrow range of 200 nt magnifies the individual areas of microRNA (10 to 40 nt) and small RNA (40 to 200 nt) allowing for high separation resolution and detailed analysis of each region.
Caution: For research use only. Not for use in diagnostic procedures.