Genomic DNA Kits
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
DNF-467-0500 DNF-468-0500 DNF-270-0240 DNF-270-0500 DNF-488-1000
CA76645-228EA 2597.65 CAD
CA76645-228 CA76645-106 CA76645-104 CA76645-460 CA76645-290 CA76645-374
Genomic DNA Kits
Electrophoresis Systems Capillary Electrophoresis Systems
Genomic DNA quality control of whole genome libraries, including sizing of gDNA extractions.
  • Broad concentration range - The HS gDNA 50 kb kit has a concentration range of 0.3 to 12 ng/µl, while the gDNA 50 kb kit covers 25 to 250 ng/µl
  • High and standard sensitivity kits - Minimize sample dilutions and choose the kit that has the right input concentration range for your needs
  • Reliable Sizing - Sizing range from 75 bp to 60 kb ensures accurate and precise sizing of gDNA samples
  • Low sample volume - Only 1 to 2 µl of sample is required, minimizing sample loss
More Product Information