TapeStation Parts and Accessories for 2200
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-5601 5067-5152 5067-5153 5067-5154 5067-5155
CA76645-032EA 513.33 CAD
CA76645-032 CA76645-134 CA76645-458 CA76645-210 CA76645-086 CA76645-036 CA76645-284
TapeStation Parts and Accessories for 2200
Electrophoresis Systems Capillary Electrophoresis Systems
Accessories and spare parts for the 2200 TapeStation system like pates and foil seals, loading tips, TapeStation test tape, needle cartridge.
  • 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
  • Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the 2200 TapeStation system
  • TapeStation test tape is available for troubleshooting and running system diagnostics tests. The Tapes are reusable and expire after two years
Our portfolio of 2200 TapeStationMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable