Bioanalyzer Small RNA Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-1549 5067-1550 5067-1548
CA76645-240EA 1033.09 CAD
CA76645-240 CA76645-020 CA76645-200
Bioanalyzer Small RNA Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
High resolution small RNA electrophoresis of fragments from 6 to 150 nt.
  • High resolution analysis, with a sizing range from 6 to 150 nucleotide, allows for separation of miRNA smears from tRNA
  • Minimal sample consumption, only use as little as 50 pg of purified miRNA or 10 ng of total RNA for analysis in 1 μl volume
  • A faster and more sensitive alternative to agarose and polyacrylamide gels analysis for detection of small RNA
  • Compatibility with multiple sample types enables analysis of total RNA, fractionated, or synthetic samples
Order Now
SPECIFICATIONS
Environmentally Preferable