DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-5589 5067-5590 5067-5602 5067-5582 5067-5583 5067-5586 5067-5588
CA76645-258EA 166.63 CAD
CA76645-258 CA76645-112 CA76645-484 CA76645-180 CA76645-488 CA76645-378 CA76645-476
DNA ScreenTape Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
DNA fragment or library analysis for sample quality control of samples up to 5000 bp.
  • A broad size range allows analysis of DNA fragments and libraries from 35 to 5000 base pairs
  • Fast results are obtained, with data delivered in approximately 1 minute per sample
  • Minimal sample consumption is required as only 1 µl of DNA sample is needed
  • DNA sample detection is possible down to 0.1 ng/µl
  • Ready-to-use ScreenTape devices, pre-packaged reagents, and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • No cross-contamination or sample carryover is caused due to the use of single lanes and individual loading tips
DNA ScreenTape analysis provides a fast, automated, and flexible DNA electrophoresis solution for sample quality control, e.g in next-generation sequencing workflows. The DNA ScreenTape assays can separate DNA fragments and libraries up to 5000 base pairs in 1 to 2 minutes/sample.
Data output yields objective quantitative and purity measurements from only 1 µl of sample. The ready-to-use ScreenTape technology makes reproducible DNA sizing and quantification possible.
Assays are compatible with all agilent TapeStation instruments.
Note: Some products within these assays contain SYBR™ green I, which is licensed from life technologies corporation for use in research and development only. SYBR™ is a registered trademark of life technologies corporation.
Caution: For research use only. Not for use in diagnostic procedures.