Bioanalyzer Parts and Accessories
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5065-4401 5065-4413 5065-9951
CA76645-350EA 2160.74 CAD
CA76645-350 CA76645-204 CA76645-160
Bioanalyzer Parts and Accessories
Electrophoresis Systems Capillary Electrophoresis Systems
Bioanalyzer spare parts, including electrode cartridges, priming stations, filters, and fuses
  • Removable electrode cartridges allow labs to buy additional cartridges dedicated for specific applications
  • Replacement test chip kits are available for troubleshooting and running diagnostic tests
  • The chips are reusable and expire after three years
  • Multiple chip priming stations are available and may be useful for core facilities or labs with multiple Bioanalyzer users
  • To ensure proper maintenance of the Bioanalyzer system additional gaskets are available in the
More Product Information