RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-5576 5067-5577 5067-5578
CA76645-246EA 371.43 CAD
CA76645-246 CA76645-294 CA76645-006
RNA ScreenTape Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
Reliable, efficient, and objective measurements of quality and quantity of RNA samples.
  • Utilizes RINe - a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Provides fast results with data delivered in approximately 1 minute per sample
  • Minimal sample consumption is required as only 1 µl of sample is needed for accurate data
  • RNA sample detection down to 5 ng/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR
More Product Information