High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-5579 5067-5580 5067-5581
CA76645-182EA 442.01 CAD
CA76645-182 CA76645-260 CA76645-286
High Sensitivity RNA ScreenTape Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
Reliable, reproducible, and objective measurements of RNA samples of limited abundance.
  • Utilizes RINe – a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Fast results are obtained in approximately 2 minutes per sample
  • Minimal sample consumption is required as only 2 µl of sample is needed
  • Detection of RNA sample with limited abundance down to 100 pg/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR
More Product Information