VWR® Performance Flammable Material Storage Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-708 76644-712 76644-716
76644-708EA 14532.95 CAD
76644-708 76644-712 76644-716
VWR® Performance Flammable Material Storage Manual Defrost Freezers with Inner Doors
Freezers Flammable Material Freezers
The inner doors on these flammable storage freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
Our Performance Series includes premium additions such as state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-708, 76644-712, and 76644-716
Ordering information: All units come with a three-year parts and labor warranty and a five-year compressor parts warranty.