VWR® Plus Flammable Material Storage Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-706 76644-710 76644-714
76644-706EA 11996.48 CAD
76644-706 76644-710 76644-714
VWR® Plus Flammable Material Storage Manual Defrost Freezers with Inner Doors
Freezers Flammable Material Freezers
The inner doors on these flammable storage freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-706, 76644-710, and 76644-714
Ordering information: All units come with a two-year parts and labor warranty and a five-year compressor parts warranty.