VWR® Plus Flammable Material Storage Refrigerators
Supplier: VWR International
VWR-HC-FRP-14P VWR-HC-FRP-17P VWR-HC-FRP-20P
76644-718EA 11918.67 CAD
76644-718 76644-722 76644-726
VWR® Plus Flammable Material Storage Refrigerators
Refrigerators
These flammable storage refrigerators offer premium cooling and protection for flammable substances.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage refrigerators meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-722
Ordering information: All units come with a two-year parts and labor warranty and a five-year compressor parts warranty.