VWR® Plus Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-730 76644-734 76644-738
76644-730EA 11435.85 CAD
76644-730 76644-734 76644-738
VWR® Plus Manual Defrost Freezers with Inner Doors
Freezers Laboratory / General Purpose Freezers
The inner doors on these Plus Series freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energyMore Product Information
Order Now
SPECIFICATIONS
Model 76644-730 76644-734 76644-738
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Defrost Manual Manual Manual
Shelves/drawers 7 inner doors 7 inner doors 7 inner doors
Door type 1 solid door, right hinged 1 solid door, right hinged 1 solid door, right hinged
Environmental certification Natural hydrocarbon refrigerants Natural hydrocarbon refrigerants Natural hydrocarbon refrigerants
Weight 90 kg (199 lbs.) 113 kg (249 lbs.) 118 kg (261 lbs.)