VWR® Performance Manual Defrost Freezers with Inner Doors
Supplier: VWR International
76644-732 76644-736 76644-740
76644-732EA 13973.05 CAD
76644-732 76644-736 76644-740
VWR® Performance Manual Defrost Freezers with Inner Doors
Freezers Laboratory / General Purpose Freezers
The inner doors on these Performance Series freezers protect stored substances with superior cooling retention.
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Performance Series showcases our most elite features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-732, 76644-736, and 76644-740
Ordering information: All units come with a three-year parts and labor warranty and a five-year compressor parts warranty.