ChemTrap® H402 Chemical Filtration Option for Safety Cabinets, Erlab
Supplier: Erlab
F2.AS F2.BE CHEMTRAPH402MC
10812-238EA 455.04 CAD
10812-238 10813-212 76640-766
ChemTrap® H402 Chemical Filtration Option for Safety Cabinets, Erlab
Air Purifiers
Add on filtration system that will filter the interior space of a flammable, acid, or corrosive cabinet.
  • User protection while saving energy
  • Easy installation, connection via bung port - no ductwork needed
  • Eliminates toxic fumes and odors while purifying laboratory air
  • Adaptable to any cabinet
  • Horizontal (H) model
The filters will capture and retain toxic fumes present within the cabinet, and will return clean air back to the laboratory, eliminating odors when the door is opened. Designed as a standalone system with no need for ducting to the outside,More Product Information