Phosphate Chemical Test Kit
Supplier: HANNA INSTRUMENTS CANADA CA
HI3833 HI3833
CA470105-388KT 35.39 CAD
CA470105-388 470105-388
Phosphate Chemical Test Kit
Environmental Test Kits
This chemical test kit that uses the ascorbic acid method to measure phosphate. It is supplied complete with all of the reagents and equipment necessary to perform approximately 50 tests.
  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Color comparison cube for simple determination
  • Complete setup-All required materials are included with the test kit, such as the sample beaker, color comparison cube, and reagent dropper bottles.
Phosphates are present in a number of products that are used by humans everyday. Some examples of the effects of phosphates are enhancing the flavor and tartness of cola drinks, as a buffering agent in controlling pH in antifreeze and delaying darkening of cut potatoes used in making french fries. Phosphates are also extensively used in detergents and cleaning fluids because of their ability to soften water and remove soil deposits.
Phosphates are particularly important for the growth and development of plant roots, and hence are one of the most common fertilizers used in agriculture. However, high concentrations of phosphates in agricultural runoff can cause environmental pollution, as they are a primary cause of eutrophication. Local laws govern the use of phosphates and the discharge levels into streams.T26.
Ordering information: Replacement reagents can be ordered.