Total Chlorine Colorimeter - Checker® HC
Supplier: HANNA INSTRUMENTS CANADA CA
HI711 HI711
CA470018-042EA 90.69 CAD
CA470018-042 470018-042
Total Chlorine Colorimeter - Checker® HC
Environmental Test Kits
Use our simple handheld colorimeter to get higher accuracy water testing results. Forget about the struggle of matching your sample to a color chart, our total chlorine colorimeter eliminates the struggle and provides you a digital total chlorine result directly. Our ammonia colorimeter brings you more precise technology than your typical chemical test kit giving you accurate and consistent results.
  • Easily carry our handheld colorimeter in your pocket
  • With one-button operation getting your total chlorine result is simple
  • Make sure your results are consistent with our built-in timer
  • Easier to use and more accurate than chemical test kits
  • Dedicated to a single parameter
  • Small size, big convenience
  • Ideal for: Water quality, education, pool and spa
The total chlorine handheld colorimeter provides a simple, accurate and cost effective way to measure total chlorine. Chlorine is a popular disinfectant used in municipal water, fruits and vegetables, as well as equipment in the food and beverage industry. Though beneficial in destroying pathogens, levels greater than 4 parts per million (ppm) is harmful to humans and therefore, must be monitored. Designed as a more accurate alternative to chemical test kits, the colorimeter provides quick, accurate results in a few easy steps.
Ordering information: Reagents will need to be repurchased. Glass cuvettes can be replaced as well. Can also buy a calibration set separately to validate meter is working correctly.