Dissolved Oxygen Chemical Test Kit
Supplier: HANNA INSTRUMENTS CANADA CA
HI3810 HI3810
CA470006-744KT 115.79 CAD
CA470006-744 470006-744
Dissolved Oxygen Chemical Test Kit
Environmental Test Kits
This is a chemical test kit that measures dissolved oxygen by modified Winkler titration. The HI3810 is supplied complete with all of the reagents and equipment necessary to perform approximately 110 tests.
  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Manual titration performed with starch indicator
  • All required materials are included with the test kit, such as the glass stoppered bottle, indicator and reagent bottles, and calibrated syringe
This test kit is a titration-based chemical test kit that determines the dissolved oxygen concentration within the 0 to 10 mg/L Ol₂ range. The test kti is supplied with all of the necessary reagents and equipment to perform the analysis. The test kit contains enough reagents for perform approximately 110 tests.The concentration of dissolved oxygen in water is extremely important in nature as well in man’s environment. In oceans, lakes, rivers, and other surface water bodies, dissolved oxygen is essential to the growth and development of aquatic life. Without oxygen, water can become toxic due to the anaerobic decaying of organic matter. In man’s environment, water must contain at least 2 mg/L of oxygen to protect water pipes from corrosion. However, boiler system water, in many cases, cannot contain greater than 10 mg/L oxygen.
Ordering information: The HI3810-100 can be ordered to replace the reagents supplied with the kit.