PUREGRIP® HPLC Solvent Reservoir Bottles GL45, Foxx Life Sciences
Supplier: Foxx Life Sciences
45R-4600-FLS 45R-4601-FLS 45R-4602-FLS 45R-4603-FLS 45R-4604-FLS 45R-5600-FLS 45R-5601-FLS 45R-5602-FLS 45R-5603-FLS 45R-5604-FLS 45R-6600-FLS 45R-6601-FLS 45R-6602-FLS 45R-6603-FLS 45R-6604-FLS 45V-4600-FLS 45V-4601-FLS 45V-4602-FLS 45V-4603-FLS 45V-4604-FLS 45V-5600-FLS 45V-5601-FLS 45V-5602-FLS 45V-5603-FLS 45V-5604-FLS 45V-6600-FLS 45V-6601-FLS 45V-6602-FLS 45V-6603-FLS 45V-6604-FLS
76379-788EA 522.27 CAD
76379-788 76379-790 76379-792 76379-794 76379-796 76379-798 76379-800 76379-802 76379-804 76379-806 76379-808 76379-810 76379-812 76379-814 76379-816 76379-818 76379-820 76379-822 76379-824 76379-826 76379-828 76379-830 76379-832 76379-834 76379-836 76379-838 76379-840 76379-842 76379-844 76379-846
PUREGRIP® HPLC Solvent Reservoir Bottles GL45, Foxx Life Sciences
Bottles HPLC Bottles
Foxx Life Sciences PUREGRIP® HPLC Reservoir Caps and Bottle Assemblies offer the safest and most ergonomic lab safety solution in the industry. These assemblies come with VersaCap Technology, A Class VI PTFE Manifold and an Air Inlet Valve combined with a Foxx 25 mm, 0.2 µm PTFE Filter for safe vapor filtration.
The Air Inlet Valve and PTFE Filter block out all harmful solvent vapors while blocking any incoming contaminants from the incoming air. This valve threads directly into any of Foxx Life Sciences HPLC Reservoir Manifolds.
The sealed VersaCap design that Foxx offers eliminatesMore Product Information