Ward's® Microfossil Slides
Supplier: Shiv Dial Sud & Sons
18-60C 18-60C1
470348-082CS 729 CAD
470348-082 470348-084
Ward's® Microfossil Slides
Slides Prepared Slides
Perfect mount to observe microfossils under the microscope.
  • Aluminum sides to ensure slides stay place
  • Glass cover slip to protect microfossils
  • Round or square cavity options
  • Packs of 100