Sartoclear® Dynamics Lab V Filtration Kits, Sartorius
Supplier: Sartorius
SDLV015005E02 SDLV025005E02 SDLV025010E02 SDLV050005E02 SDLV050010E02 SDLV050020E0E SDLV100010E02 SDLV100020E0E SDLV100040E0E SDLV015002E0E SDLV050005E02 SDLV100020E0E SDLV050010E02 SDLV025010E02 SDLV100040E0E
76595-334PK 540.77 CAD
76595-334 76595-362 76595-340 76595-318 76595-316 76595-320 76595-354 76595-348 76595-324 76595-332 CA76595-318 CA76595-348 CA76595-316 CA76595-340 CA76595-324
Sartoclear® Dynamics Lab V Filtration Kits, Sartorius
Filters Bottle Top Filters
Sartoclear Dynamics® Lab V kits enable clarification and sterile filtration to be performed in a single step.
  • Clarify high-density mammalian cell culture in minutes
  • Out-of-the-box solution
  • Fast and effortless
  • No need for centrifugation
These kits simplify the process by fully eliminating the centrifugation step otherwise needed for clarification. As a result, they enable harvested cell cultures to be efficiently clarified and sterilized in minutes – quickly and easily.
Sartoclear Dynamics® Lab V kits are designed for simultaneous clarification andMore Product Information