Sartolab® Storage Bottles, Single-Use, Polystyrene
Supplier: Sartorius
18022E 18023E 18024E 18025E 18024E 18022E 18025E 18023E
76595-306PK 79.08 CAD
76595-306 76595-326 76595-310 76595-312 CA76595-310 CA76595-306 CA76595-312 CA76595-326
Sartolab® Storage Bottles, Single-Use, Polystyrene
Bottles Plastic Bottles
Single-use polystyrene bottles for storage of media, buffers and other aqueous solutions.
  • Low-profile design for easy gripping with one hand
  • Ergonomically designed cap to provide a rigid grip and one-hand handling of each bottle
  • The bottles with an indented bottom are stackable to save space in the refrigerator and in the freezer
  • Caps (45 mm) with airtight seal to minimize the risk of contamination
  • Graduation marks on the bottles ensure accuracy and easy reading
  • Designated writing field for clear labeling of samples
  • The materials used in the production are medical-grade for the highest quality, without any animal origin
  • Sterile and endotoxin-free
Sartorius single-use polystyrene (PS) storage bottles are designed for safe, secure storage of tissue culture media and sera, buffers and other aqueous solutions. The bottles can be used with all Sartolab® RF / BT vacuum filtration units with a 45 mm neck size.