Exo-Terra® Screen Terrarium
Supplier: ROLF C. HAGEN (USA) CORP.
Exo-Terra®
PT2677 PT2679 PT2678 PT2676
470347-684EA 356.05 CAD
470347-684 470347-708 470347-844 470347-752
Exo-Terra® Screen Terrarium
Animal Supplies Animal Cages
The Exo-Terra® Screen Terrarium is a perfect habitat for arboreal reptiles and amphibians that are sensitive to stagnant air.
  • Excellent non-restricted airflow provides optimal ventilation while minimizing odors and fungus growth in the reptile's habitat
  • Screen design provides a broader temperature gradient, which helps reptiles to regulate their body temperature more effectively and prevents overheating
  • Aluminum screen mesh allows ultraviolet rays from UVB lights or the sun to penetrate deep inside the enclosure
  • Substrate tray allows to place a small layer of substrate to absorb spray water and increase humidity levels; it can be removed easily from the enclosure through the swiveling bottom door
  • Large hinged front door(s)
  • Sturdy nickel-plated latches add stability to the enclosure and keep reptiles safe and secure
Ideal for chameleons and other tree dwellers
Ordering information: Includes substrate tray.