D-(+)-Sucrose, Molecular Biology Grade, Calbiochem®, Millipore®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

573113-5KG 573113-0.5KG
CA80503-442EA 571 CAD
CA80503-442 CA95061-670
D-(+)-Sucrose, Molecular Biology Grade, Calbiochem®, Millipore®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR