Sodium dihydrogen phosphate monohydrate, Molecular Biology Grade

Supplier: MilliporeSigma

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

567549-1KG 567549-250GM
CA80503-416EA 525.23 CAD
CA80503-416 CA80503-418
Sodium dihydrogen phosphate monohydrate, Molecular Biology Grade
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
MDL Number: MFCD00149208
CAS Number: 10049-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR