Dipotassium hydrogen orthophosphate trihydrate, Molecular Biology Grade

Supplier: MilliporeSigma

Synonyms: Potassium hydrogen phosphate trihydratePotassium phosphate dibasic trihydrateDipotassium hydrogen phosphate trihydratedi-Potassium hydrogen orthophosphate trihydratedi-Potassium hydrogen phosphate trihydrate

529567-1KG 529567-1KG
CA80503-246EA 514.06 CAD
80503-246 CA80503-246
Dipotassium hydrogen orthophosphate trihydrate, Molecular Biology Grade
di-Potassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
MDL Number: MFCD00149926
CAS Number: 16788-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR