Dithiothreitol (DTT, Cleland's reagent), Molecular Biology Grade

Supplier: MilliporeSigma

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

233156-10GM 233156-1GM
CA80502-176EA 819.91 CAD
CA80502-176 CA80502-178
Dithiothreitol (DTT, Cleland's reagent), Molecular Biology Grade
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
MDL Number: MFCD00004877
CAS Number: 3483-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR