(±)-10-Hydroxycamptothecin (from Camptotheca acuminata)

Supplier: MilliporeSigma

Synonyms: 10-HydroxycamptothecinNSC1071241-HCPT4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

390238-25MG 390238-25MG
CA80502-006EA 538.76 CAD
80502-006 CA80502-006
(±)-10-Hydroxycamptothecin (from Camptotheca acuminata)
(±)-10-Hydroxycamptothecin
Formula: C₂₀H₁₆N₂O₅
MW: 364.35 g/mol
MDL Number: MFCD09027971
CAS Number: 64439-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR