(4-Nitrophenyl)-α-D-glucopyranoside

Supplier: MilliporeSigma

Synonyms: 4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside

487506-1GM 487506-5GM
CA80110-004EA 157.56 CAD
CA80110-004 CA80110-006
(4-Nitrophenyl)-α-D-glucopyranoside
(4-Nitrophenyl) α-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
MDL Number: MFCD00064088
CAS Number: 3767-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR