Guanidinium hydrochloride

Supplier: MilliporeSigma

369079-1KG 369079-5KG
CA80108-144EA 758.86 CAD
CA80108-144 CA80108-146
Guanidinium hydrochloride
Guanidine hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MDL Number: MFCD00013026
CAS Number: 50-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR