3-10 Detergent, Zwittergent®
Supplier: MilliporeSigma
Synonyms:
3-(Decyldimethylazaniumyl)propane-1-sulfonate, 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate, 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulfonate
693021-25GM
693021-5GM
CA80058-852EA
1169.79
CAD
CA80058-852
CA80058-854
3-10 Detergent, Zwittergent®
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10)
Formula:
C₁₅H₃₃NO₃S MW: 307.4924 g/mol |
MDL Number:
MFCD00036908 CAS Number: 15163-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...