Tauroursodeoxycholate sodium ≥97% (by TLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma

580549-1GM 580549-5GM
CA80058-720EA 408.03 CAD
CA80058-720 CA80058-722
Tauroursodeoxycholate sodium ≥97% (by TLC), Calbiochem®, Millipore®
Tauroursodeoxycholate sodium

Tauroursodeoxycholic is a detergent useful for the solubilization of lipids and membrane-bound proteins.

Formula: C₂₆H₄₅NNaO₆S
MDL Number: MFCD00065451
CAS Number: 35807-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR