D-(-)-Sorbitol

Supplier: MilliporeSigma

Synonyms: D-SorbitolSorbitolD(-)-Glucitol(Sorbitol)(2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol

56755-1KG 56755-1KG
CA80058-652EA 245.78 CAD
CA80058-652 80058-652
D-(-)-Sorbitol
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
MDL Number: MFCD00004708
CAS Number: 50-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR