N-Octyl-β-D-glucopyranoside, ULTROL®

Supplier: MilliporeSigma

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

494460-1GM 494460-250MG 494460-5GM
CA80058-522EA 728.88 CAD
CA80058-522 CA80058-524 CA80058-526
N-Octyl-β-D-glucopyranoside, ULTROL®
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR