3-(Benzyldimethylammonio)propanesulfonate
Supplier: MilliporeSigma
Synonyms:
NDSB-256, 3-[dimethylbenzylamino]propanesulfonic acid, 3-(Benzyldimethylammonio) propane-1-sulphonate, 3-(Benzyldimethylammonio) propane-1-sulfonate, 3-[dimethylbenzylamino]propanesulphonic acid
480010-25GM
480010-5GM
CA80058-460EA
1268.13
CAD
CA80058-460
CA80058-462
3-(Benzyldimethylammonio)propanesulfonate
3-(Benzyldimethylammonio)propanesulfonate
Formula:
C₁₂H₁₉NO₃S MW: 257.348 g/mol |
MDL Number:
MFCD00225018 CAS Number: 81239-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...