NDSB-195

Supplier: MilliporeSigma

Synonyms: Ethyl(dimethylaminio)propane sulphonate, 3-(Ethyldimethylaminio)propane-1-sulfonate, 3-(Ethyldimethylammonio)propane-1-sulfonate, 3-(Ethyldimethylaminio)propane-1-sulphonate, 3-(Ethyldimethylammonio)propane-1-sulphonate, NDSB 195, Dimethylethylammonium propane sulfonate, Ethyl(dimethylaminio)propane sulfonate, Dimethylethylammonium propane sulphonate

480001-25GM 480001-5GM
CA80058-452EA 1116.62 CAD
CA80058-452 CA80058-454
NDSB-195
NDSB-195
Formula: C₇H₁₇NO₃S
MW: 195.277 g/mol
MDL Number: MFCD01319124
CAS Number: 160255-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR