Magnesium chloride hexahydrate
Supplier: MilliporeSigma
Synonyms:
Magnesium dichloride
442615-500GM
442615-500GM
CA80058-330EA
200.19
CAD
CA80058-330
80058-330
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Formula:
MgCl₂·6H₂O MW: 203.3 g/mol |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...