Tricine, ULTROL®

Supplier: MilliporeSigma

39468-100GM 39468-1KG
CA80058-264EA 232.25 CAD
CA80058-264 CA80058-266
Tricine, ULTROL®
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
MDL Number: MFCD00004277
CAS Number: 5704-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR