2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid free acid, ULTROL®

Supplier: MilliporeSigma

Synonyms: BES, N,N-Bis-(2-hydroxyethyl)-2-ethanesulfonic acid, 2-(Bis(2-hydroxyethyl)amino)ethanesulfonic acid, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid

391334-100GM 391334-25GM
CA80058-216EA 776.85 CAD
CA80058-216 CA80058-218
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid free acid, ULTROL®
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formula: C₆H₁₅NO₅S
MDL Number: MFCD00007533
CAS Number: 10191-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR