HEPES sodium salt, ULTROL®
Supplier: MilliporeSigma
Synonyms:
Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt, HEPES sodium, Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate, HEPES Na
Formula:
C₈H₁₇N₂NaO₄S MW: 260.29 g/mol |
MDL Number:
MFCD00036463 CAS Number: 75277-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...