Glycine

Supplier: MilliporeSigma

Synonyms: Aminoacetic acidH-Gly-OHGlycocoll

3570-500GM 3570-1KG 3570-500GM 3570-1KG
CA80058-142EA 177.16 CAD
CA80058-142 CA80058-140 80058-142 80058-140
Glycine
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
MDL Number: MFCD00008131
CAS Number: 56-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR