Sodium deoxycholate, ULTROL®
Supplier: MilliporeSigma
Synonyms:
Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
264103-25GM
264103-5GM
CA80058-008EA
1212.54
CAD
CA80058-008
CA80058-010
Sodium deoxycholate, ULTROL®
Sodium deoxycholate
Formula:
C₂₄H₃₉NaO₄ MW: 414.56 g/mol |
MDL Number:
MFCD00064139 CAS Number: 302-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...