Sodium cholate

Supplier: MilliporeSigma

229101-250GM 229101-50GM
CA80057-936EA 744.47 CAD
CA80057-936 CA80057-938
Sodium cholate
Sodium cholate
Formula: C₂₄H₃₉NaO₅
MDL Number: MFCD00150749
CAS Number: 361-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR