3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), Molecular Biology Grade

Supplier: MilliporeSigma

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS

220201-10GM 220201-1GM 220201-25GM 220201-5GM
CA80057-914EA 750.67 CAD
CA80057-914 CA80057-916 CA80057-918 CA80057-920
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), Molecular Biology Grade
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 13,02052

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR